GE Healthcare Pharmalyte™ Carrier Ampholytes Available on GSA/VA Contract for Federal Government customers only.

Use in Isoelectric focusing. GE Healthcare Life Sciences™ Pharmalyte™ Carrier Ampholytes prepared by the co-polymerization of glycine, glycylglycine, amines, and epichlorohydrin, are available in five broad-range and four narrow-range pH intervals.

Manufacturer: GE Healthcare 17056201

 View more versions of this product

Catalog No. 45-000-102

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) Used as carrier for high resolution isoelectric focusing, for intervals for special dedicated applications in Polyacrylamide Gels
    Format Liquid
    Formulation Co-polymerization of Glycine, Glycylglycine, Amines and Epichlorohydrin
    Product Name Pharmalyte carrier Ampholyte
    Quantity 25mL
    Storage Requirements 2° to 8°C
    Color Colorless to Light Yellow
    For Use With (Equipment) Precast PhastGel™ Gels for PhastSystem
    pH 4.2 to 4.9

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit