GE Healthcare Pharmalyte™ Carrier Ampholytes Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: GE Healthcare 17045201

Catalog No. 45-000-075

 N/A
N/A

This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Format Liquid
    Formulation Co-polymerization of Glycine, Glycylglycine, Amines and Epichlorohydrin
    Product Name Pharmalyte carrier Ampholyte
    Quantity 25mL
    Storage Requirements 2° to 8°C
    Color Colorless to Light Yellow
    For Use With (Application) Used as carrier for high resolution isoelectric focusing, can be used directly for Agarose, Ultrodex or Polyacrylamide Gels and form linear gradients
    For Use With (Equipment) Precast PhastGel™ Gels for PhastSystem
    pH 4 to 6.5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit