Learn More
GE Healthcare PhastGel™ Buffer Strips Available on GSA/VA Contract for Federal Government customers only.
Pre-prepared 3% agarose blocks containing buffer, for use in conjunction with PhastGel gradient and homogeneous gel media to generate a multiphasic buffer system during electrophoresis
Manufacturer:  GE Healthcare 17051601
 View more versions of this product
Catalog No. 45-000-089
Add to cart
Description
Description
  • PhastGel Buffer Strips are available in three types – PhastGel DNA, PhastGel SDS, and PhastGel Native
  • Select from homogeneous and gradient polyacrylamide gels.
  • Save time and improve reproducibility with precast gels, buffer strips, and staining kits.
  • Achieve detection sensitivity comparable to that of radioisotopes with ready-to-use silver staining kits
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
0.2M Tricine, 0.2M Tris, 0.55% SDS, pH 8.1
Used for industrial applications such as analytical chemistry and research
Solid
PhastGel™ Buffer strips-SDS
4° to 8°C
Colorless
PhastGel™ on PhastSystem automated flatbed electrophoresis units
N-(Tri(Hydroxymethyl)Methyl)Glycine 1 to 5%, Trometamol 1 to 5%
20 Strips
10 PhastGel™