Learn More
GE Healthcare PhastGel™ Buffer Strips
Pre-prepared 3% agarose blocks containing buffer, for use in conjunction with PhastGel gradient and homogeneous gel media to generate a multiphasic buffer system during electrophoresis
Manufacturer:  GE Healthcare 17051701
 View more versions of this product
Catalog No. 45-000-090
Add to cart
Description
Description
  • Ready made buffer strips eliminate the need to prepare electrophoresis buffers
  • Supplied as 10X pairs - for both cathode and anode
  • Choice of Native or Denaturing (SDS) buffer systems
Specifications
Specifications
0.88M L-alanine, 0.25M Tris, pH 8.8
Used for industrial applications such as analytical chemistry and research
Solid
PhastGel™ Buffer strips-native
4° to 8°C
Colorless
PhastGel™ on PhastSystem automated flatbed electrophoresis units
Trometamol 1 to 5%
20 Strips
10 PhastGel™