Learn More
GE Healthcare PhastGel™ Precast Gels Available on GSA/VA Contract for Federal Government customers only.
Select from homogeneous and gradient polyacrylamide gels. GE Healthcare PhastGel™ Precast Gels save time and improve reproducibility.
Manufacturer:  GE Healthcare 17062201
 View more versions of this product
Catalog No. 45-000-119
Add to cart
Description
Description
  • Select from homogeneous and gradient polyacrylamide gels
  • Separate proteins and peptides on denaturing and native
  • Save time and improve reproducibility using precast gels with native or denaturing buffer strips and staining kits, buffer strips, and staining kits
  • Achieve detection sensitivity comparable to that of radioisotopes with ready-to-use silver staining kits
  • Gels are cast on a plastic backing for ease of handling
PhastGel homogenous gels
  • Available in 7.5%, 12.5%, and 20% polyacrylamide with stacking gel, for flexibility in separation, extending the resolution between proteins within a narrow MW range
  • A high density PhastGel (20%) is also available containing PEG to optimise separations below Mr 20000 increasing resolution
PhastGel gradient gels
  • Available in concentration gradients of 4-15%,10-15% and 8-25% with stacking gel, for separation of complex mixtures, giving a good overall view of total composition within a sample
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
125°C
Used for industrial applications such as analytical chemistry and research
4.9 cm
82° to 86°C
5% T/3% C
0.017 in.
5 cm
GelBond polyester film
7.5%
Denaturing, Native
Protein
PhastGel Homogeneous – 7.5
1.92 in.
4.9 cm
71.08
4 - 8°C
1.9 in.
Precast Gel
70 to 230 Mr
Tris-Acetate
0.45 mm
10 Pack