GE Healthcare PhastGel™ Precast Gels Available on GSA/VA Contract for Federal Government customers only.

Select from homogeneous and gradient polyacrylamide gels. GE Healthcare Life Sciences™ PhastGel™ Precast Gels save time and improve reproducibility.

Manufacturer: GE Healthcare 17054001

 View more versions of this product

Catalog No. 45-000-094

  Check Availability
Add to Cart

    Specifications

    Material Polyacrylamide,GelBond™ Polyester Film
    Boiling Point 125°C
    Height (English) 1.69 in.
    Height (Metric) 4.3cm
    Buffer 112mM Acetate, 112mM Tris, pH 6.4
    Color Colorless,Colorless
    Concentration 10 to 15%
    Melting Temperature 82° to 86°C
    Molecular Weight 71.08
    Separation Range 80 to 440Mr
    Stacking Zone 6% T/3% C
    Storage Requirements 4° to 8°C
    Thickness (English) 0.017 in.,0.017 in.
    Thickness (Metric) 0.04cm
    Width (English) Gel 1.9 in.
    Width (Metric) Gel 5cm,5cm
    For Use With (Application) Industrial applications such as analytical chemistry and research
    Format Precast Gel
    Formulation Continuous Acrylamide gradients in the range 10 to 15%

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit