Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare PhastGel™ Precast Gels Available on GSA/VA Contract for Federal Government customers only.

Select from homogeneous and gradient polyacrylamide gels. GE Healthcare PhastGel™ Precast Gels save time and improve reproducibility.

Manufacturer:  GE Healthcare 17067901

 View more versions of this product

Catalog No. 45-000-129


  Check Availability
Add to cart

Specifications

Description

  • Select from homogeneous and gradient polyacrylamide gels
  • Separate proteins and peptides on denaturing, native, and isoelectric focusing gels
  • Save time and improve reproducibility using precast gels with native or denaturing buffer strips and staining kits, buffer strips, and staining kits
  • Achieve detection sensitivity comparable to that of radioisotopes with ready-to-use silver staining kits
  • Gels are cast on a plastic backing for ease of handling

  • PhastGel homogenous gels
    • Available in 7.5%, 12.5%, and 20% polyacrylamide with stacking gel, for flexibility in separation, extending the resolution between proteins within a narrow MW range
    • A high density PhastGel (20%) is also available containing PEG to optimise separations below Mr 20000 increasing resolution
    PhastGel homogenous gels
    • Available in concentration gradients of 4-15%,10-15% and 8-25% with stacking gel, for separation of complex mixtures, giving a good overall view of total composition within a sample
  • PhastGel IEF gels
    • Homogeneous gels in pH ranges of 3-9, 5-8 and 4-6.5, for separations of proteins by isoelectric point

    Description & Specifications

    Specifications

    125°C
    Colorless
    High-density, 20% Polyacrylamide, 30% Ethylene Glycol
    4.3cm
    20 to 60Mr
    0.017 in.
    1.9 in.
    Polyacrylamide
    112mM Acetate, 112mM Tris, 30% Ethylene Glycol, pH 6.4
    Industrial applications such as analytical chemistry and research
    1.69 in.
    82° to 86°C
    4° to 8°C
    0.04cm
    5cm
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit