Learn More
GE Healthcare pI Markers
Run these markers in parallel with the experimental samples on isoelectric focusing gels. GE Healthcare Life Sciences™ pI Markers are lyophilized mixtures of stable, salt-free, highly purified proteins.
Manufacturer:  GE Healthcare 17047201
 View more versions of this product
Catalog No. 45-000-082
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
The markers may be run in parallel with the experimental samples on isoelectric focusing gels and are not prestained.
  • The precisely defined pI of the markers allows a permanent and accurate calibration of sample components
  • pH gradients and pI can be measured to within 0.01 to 0.05 pH unit
  • The progress of the focusing experiment can be monitored visually by the focusing of methyl red or Cytochrome c
Not for use in denaturing (urea) systems.
Specifications
Specifications
1 Kit: 10 Vials
To be used for isoelectric focusing (IEF)
Pepsinogen 2.80 pI (Native), Amylglucosidase 3.50 pI (Native), Methyl red (dye) 3.75 pI (Native), Glucose Oxidase 4.25 pI (Native), Trypsin inhibitor 4.55 pI (Native), bLactoglobulin A 5.20 pI (Native), Carbonic Anhydrase B (Bovine) 5.85 pI (Native)
4°C
White
Lyophilized
2.5 to 6.5
pl Marker Kit
Safety and Handling
Safety and Handling
ShelfLife : 2 Years