Learn More
GE Healthcare PlusOne DNA Silver Staining Kit
Minimizes production of toxic waste and optimizes sensitivity and robustness making the kit especially suitable for use in automatic staining equipment
Manufacturer:  GE Healthcare 17600030
Catalog No. 45-000-341
Add to cart
Description
Description
  • Typical sensitivity: 20 to 50pg DNA/band
  • Reaction time: <1.5 hr.
  • Kit contents: fixing solution, staining solution, sodium carbonate solution, 37% formaldehyde, 2% sodium thiosulfate, stopping and preserving solution, instruction booklet, short instructions (laminated sheet)
Specifications
Specifications
1 Kit
Automatic staining equipment, Standard manual Silver staining of DNA in Polyacrylamide gels
Fixing solution 5X: Benzene Sulphonic Acid, 3% w/v in 24% v/v Ethanol 500mL, Staining solution 5X: Silver Nitrate, 1% w/v Benzene Sulphonic Acid, 0.35 % w/v 500mL, Sodium Carbonate solution 5X: Sodium Carbonate, 12.5% w/v 500mL, Formaldehyde 37%
10° to 30°C
DNA Silver Staining Kit provides reproducible and sensitive nonradioactive detection of nucleic acids in polyacrylamide gels in less than 1.5 h. Brings speed, convenience and very high sensitivity to the staining of proteins and nucleic acids.
Liquid
Silver Stain
DNA Silver Staining Kit