Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare PlusOne™ Reagents Available on GSA/VA Contract for Federal Government customers only.

High quality electrophoresis reagents. GE Healthcare Life Sciences™ PlusOne Reagents are a low toxicity, environmentally safe solvent for high quality electrophoresis.

Manufacturer:  GE Healthcare 17133001

 View more versions of this product

Catalog No. 45-000-243


Add to cart

Description

Description

Urea

  • Provides highest resolution and the cleanest results for IEF performed under denaturing conditions
  • Urea solubilizes and unfolds most proteins to their fully random conformation, with all ionizable groups exposed to solution
  • 8M urea included in the 2-D sample solution
Repel-Silane ES
Low toxicity and environmentally safe solvent
  • Inhibits sticking of polyacrylamide gels, agarose gels, and nucleic acids to glass surfaces
  • Bind-Silane
    • Used for covalently attach polyacrylamide gels to a glass surface
    • Firmly attaches to the glass during staining and drying procedures

    Urea:

    • Molecular Weight: 60.06
    • Physical state: Solid
    • Color: White
    • Pack of 500g
    Repel-Silane ES
    • Molecular Weight: 129.06
    • Physical state: Liquid
    • Color: Colorless
    • Pack of 500mL
    Bind-Silane
    • Molecular Weight: 248.4
    • Physical state: Liquid
    • Color: Pale
    • Pack of 25mL

    IEF, PAGE, sequencing

    This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

    Specifications

    Specifications

    25 mL
    PlusOne Bind-Silane
    Liquid
    248.4
    2530-85-0
    Pale Color
    Used in IEF, PAGE and Sequencing
    −48°C
    10-30ᵒC
    Bind-Silane
    Safety and Handling

    Safety and Handling

    ShelfLife : 3 Years

    Documents
    Provide Content Correction

    We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

    Product Title

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

    Cancel Submit