Learn More
GE Healthcare PlusOne™ Silver Staining Kit for Protein
Use for reproducible and sensitive staining of proteins in polyacrylamide gels. PlusOne™ Silver Staining Kit for Protein enables staining of proteins in less than two hours.
Manufacturer:  GE Healthcare 17115001
Catalog No. 45-000-190
Add to cart
Includes: Sodium acetate, 5% sodium thiosulfate, EDTA-Na2, sodium carbonate, 2.5% silver nitrate, 37% formaldehyde, and 25% glutaraldehyde
Description
Description
  • Typical sensitivity: 0.2 to 0.6ng protein/band
  • Use with 2-D electrophoresis and mass spectrometry by omitting glutaraldehyde from staining procedure
Order Info
Components supplied in quantities sufficient to stain 10 precast 12.5 x 26cm gels, 20 minigels (8-10 x 8cm), or 30 to 60 PhastGel™ Gels.
Specifications
Specifications
1 Kit
For use in Hoefer automated gel stainer as well as for manual silver staining. Silver staining is a highly sensitive method for visualizing proteins separated in SDS-polyacrylamide gels.
10 to 30°C
Protein Silver Staining Kit
PlusOne Silver Staining Kit, Protein and PlusOne DNA Silver Staining Kit are two silver staining kits that bring speed, convenience, and very high sensitivity to the staining of proteins and nucleic acids in polyacrylamide electrophoresis. gels.
Sodium Acetate: 10 x 17g packets, 5% Sodium Thiosulfate: 1 x 100mL bottle, EDTA-Na2: 10 x 3.65g packets, Sodium Carbonate: 10 x 6.25g packets, 2.5% Silver Nitrate: 2 x 125mL bottles, 37% Formaldehyde: 1 x 2.5mL bottle, 25% Glutaraldehyde: 1 x 12.5mL bottle
10 DALT precast gels, 10 precast 12.5 x 26cm gels, 20 mini slab gels (8 to 10 x 8cm) or 30 to 60 PhastGel™
Safety and Handling
Safety and Handling
ShelfLife : 1 Year