Learn More
GE Healthcare Polybuffer Solutions Available on GSA/VA Contract for Federal Government customers only.
Polybuffer solutions are used for seperating proteins by their isoelectric pint vial chromatofocusing, forming linear pH gradients after titration on Mono P columns.
Manufacturer:  GE Healthcare 17071401
 View more versions of this product
Catalog No. 45-000-136
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
250mL
Polybuffer 96 solution
1 to 14
4° to 8°C
Resolves pI differences of 0.04 pH units, use for pH gradients that should begin above pH 7