Learn More
GE Healthcare Precision Column Holder Available on GSA/VA Contract for Federal Government customers only.
Adapts PC columns designed for SMART System and for use with other high performance chromatography systems including AKTAdesign systems
Manufacturer:  GE Healthcare 17145501
 View more versions of this product
Catalog No. 45-000-253
Add to cart
Includes: Precision Column Holder, (Cat No. 45-000-253)
Column holder for 3-5cm columns, 1; Column holder for 30cm columns, 1; Screw caps, 4; End fittings, 4; Stop plugs, 4; Union M6 female 1/16 in. male 4
Column Holder, 10cm (Cat. No. 45-000-255)
Column holder for 10cm columns, 1; Screw caps, 2; End fittings, 2; Stop plugs, 2; Union M6 female 1/16 in. male, 2; Steering caps for steel column, 2
Description
Description
  • Low dead volume fittings maintain resolution
  • Includes all necessary tubing fittings and instructions
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Each
Classic AKTA™, Superdex™ Peptide PC standard chromato graphy systems column and 3 to 5cm column of most HPLC systems
Column holder
Stainless Steel