Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Processor Plus Blot Processing Trays

For blot processing

Manufacturer:  GE Healthcare 80644423

 View more versions of this product

Catalog No. 45-002-206


Add to cart

Includes: For Blot Processing Tray Pack
  • One tray support
  • Three blot processing mini trays
  • Three blot processing standard trays
  • One blot lid assembly with valves and tubing
  • One blot tray base
  • Four reagent bottles and a rack
  • Two wast bottle caps
  • One spirit level
  • One lid support

Description

Description

Blot Processing Mini Tray

  • Tray volume: 10 to 50mL per chamber
  • Operating temperature: 4° to 40°C
Blot Processing Tray Pack
  • Trays for four mini- (up to 9 x 9.5cm) standard sized blots
  • High-speed peristaltic pump for fast delivery and removal of reagents
  • One 10-port valve for multiple reagent protocols and hazardous waste segregation
  • One rhomboid-shaped tray with smooth and even rocking for rapid fluid mixing and coverage
  • Nine preprogrammed and four user-defined protocols for proteins and nucleic acids
  • Protocol entry and editing, up to 30 steps
  • Removable protocol key for storing and protects critical protocols
 
Specifications

Specifications

Blot Processing Tray Pack
39.2°F to 104°F
Processor Plus™ System
1 Kit
4°C to 40°C
Blot Processing Tray Pack
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit