Learn More
GE Healthcare Processor Plus™ Tubing Kits
Iintelligent fluid delivery system that handles both both Western blot processing and acrylamide gel staining
Manufacturer:  GE Healthcare 80644803
 View more versions of this product
Catalog No. 45-002-197
Add to cart
Includes:
  • 10 x Tubing (PVC) i.d. 3.18mm (0.125 in.) o.d. 4.76mm (0.19 in.) L = 300mm (24 in.)
  • 10 x Suction pipes
  • 10 x Label, marked 0-9
  • 1 x Tubing (Tygon) L = 250mm (10 in.) with barbed connector and coupling for inner pump head
  • 1 x Tubing (Tygon) L = 250mm (10 in.) with coupling
  • 1 x Tubing (Tygon) L = 51mm (2 in.)
  • 10 x Pump head dampeners
Description
Description
  • Change processing mode with a fast and simple switch of a tray
  • Programmable control of solution, volume, processing time and waste container
  • Westerns: high sensitivity chemiluminescent ECL™, ECL Plus™, or chromogenic protocols
    • Staining
    : Coomassie™ blue or high-sensitivity silver staining for both protein and nucleic acid
Specifications
Specifications
Inner pump tubing, outer pump tubing, ten dampeners
Pump Tubing
Tubing Kit