GE Healthcare Prostaglandin E2 Biotrak™ Assay

Enzyme immunoassay system suitable for plasma, urine, or cell culture supernatant samples

Manufacturer: GE Healthcare RPN22210

 View more versions of this product

Catalog No. 45-500-151

  Check Availability
Add to Cart

    Specifications

    For Use With (Application) ELISA
    Assay Range 20 to 640pg/mL
    Product Type Prostaglandin E2 Biotrak assay
    Quantity 10 x 96 wells
    Sample Type Plasma (EDTA, Citrate), Urine, Tissue Homogenates, Cell Culture, Cell Culture Supernatant
    Sensitivity 16pg/mL
    Total Assay Time 5 hr.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit