Learn More
GE Healthcare Protein A Sepharose™ 6MB Gel
For separation of cells coated with antibodies
Manufacturer:  GE Healthcare 17046901
Catalog No. 45-002-235
Add to cart
Description
Description
  • Binds Fc portion of IgG antibodies from a wide range of species
  • Binds 5mg human IgG/mL gel
  • Eliminates the need for a range of immunoadsorbent columns
  • Column may be regenerated between separations by washing with low-pH buffer
Order Info
1mg Protein A/mL gel
Specifications
Specifications
5mg Human IgG/mL
For separation of cells coated with antibodies
10mL
4° to 30°C