GE Healthcare Protein and Peptide Molecular Weight Markers Available on GSA/VA Contract for Federal Government customers only.

Run these markers to determine the molecular weight of uncharacterized protein samples with precision. GE Healthcare Life Sciences™ Protein and Peptide Molecular Weight Markers work by comparing unknowns with carefully selected standards.

Manufacturer: GE Healthcare 17061501

 View more versions of this product

Catalog No. 45-000-115

  Check Availability
Add to Cart

    Specifications

    Product Type HMW-SDS marker kit
    Quantity 10 Vials
    Kit Contents Myosin: 25μg, 220000 (Mr), rabbit muscle (source), a-2-Macroglobulin: 100μg, 170000 (Mr), bovine plasma (source), b-Galactosidase: 16μg, 116000 (Mr), E. coli (source), Transferrin
    Color Blue
    Storage Requirements 4°C
    For Use With (Application) SDS electrophoresis
    Format Solid

    • Get clear, sharp bands for easy measurement of migration distances
    • Bands can be visualized by using Coomassie™ Brilliant Blue or silver staining

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit