Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GE Healthcare RESOURCE Columns for HIC
Excellent for polishing in a purification process
Manufacturer: GE Healthcare 17118401
Description
- Prepacked columns for purification of proteins and peptides with hydrophobic interaction chromatography (HIC)
- High performance at low back pressure, even with high sample loads of up to 25mg of protein
- Ligands coupled to 15µm monodispersed rigid polystyrene/divinylbenzene matrix
- Designed for low- to medium- pressure systems ranging from peristalic pump to AKTA design and FPLC Systems
- RESOURCE PHE will have strongest hydrophobicity followed by RESOURCE ISO and RESOURCE ETH, successively
- Material of column: Polyetheretherketone (PEEK)
- Bed dimensions 6.4 x 30mm
- Maximum pressure 15 bar
- Flow rate: 0.8 to 4.8mL/min
- Storage: 4°C to 30°C, 20% Ethanol
Media
- Matrix: Polystyrene/divinylbenzene
- Stable in common buffers, chaotropic agents, detergents, and polar organic solvents
- Average particle size: 15Glc mum
- pH Stability Working Range: 2 to 12
- Flow velocity: 150cm/h - 900cm/h
Specifications
1 x 1 mL | |
10 cm | |
150 to 900 cm/hr | |
3.937 in. | |
Connectors for high- and low-pressure chromatography equipment and instructions | |
PEEK | |
15 to 15 µm | |
Polystyrene/Divinylbenzene |
3.937 in. | |
RESOURCE ETH column | |
1 mL | |
10 cm | |
Ether | |
15 bar | |
2 to 12 | |
4°C to 30°C, 20% Ethanol |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title