Learn More
GE Healthcare RESOURCE™ and SOURCE™ 15 and 5RPC columns
Designed for rapid screening experiments, method development and small-scale protein purification using reversed phase chromatography
Manufacturer:  GE Healthcare 17118101
 View more versions of this product
Catalog No. 45-002-024
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
1mL
580psi
Polystyrene/Divinyl Benzene
1 to 12
No
6.4mm
10mL/min.
Polyetheretherketone
Monosized
Reverse Phase Column
15μm
Matrix surface