GE Healthcare rmp Protein A Sepharose™ Fast Flow Affinity Media Available on GSA/VA Contract for Federal Government customers only.

Designed for high-purity separation of monoclonal and polyclonal antibodies

Manufacturer: GE Healthcare 17513802

 View more versions of this product

Catalog No. 45-002-975

  Check Availability
Add to Cart

    Specifications

    Binding Capacity >Min. 22mg Human IgG/mL
    Material Recombinant Protein A coupled with Sepharose 4 Fast Flow base matrix
    Matrix 4% Cross-Linked Agarose
    Storage Buffer 20% Ethanol
    Particle Size 60 to 165μm
    Product Type rmp Protein A Sepharose fast flow media
    For Use With (Application) For high-purity separation of monoclonal and polyclonal antibodies
    Ligand Type Recombinant Protein A (E. coli)
    Quantity 25mL
    Stability Stable in aqueous Buffers commonly used in Protein A Chromatography
    Storage Requirements 2 to 8°C
    pH Range 3 to 10

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit