Learn More
GE Healthcare Sample Applicator for PhastSystem™ Electrophoresis System
Sample applicators for PhastSystem Electrophoresis Systems
Manufacturer:  GE Healthcare 18161801
 View more versions of this product
Catalog No. 45-000-475
Add to cart
Description
Description
For use with PhastSystem Electrophoresis Systems
Specifications
Specifications
8 x 1 μL
PhastGel™ System
Sample Applicator