GE Healthcare Select-a-Dye mono-Reactive Dye Packs Available on GSA/VA Contract for Federal Government customers only.
Enables a variety of labeling strategies using NHS ester, maleimide, or hydrazide to be investigated
Manufacturer: GE Healthcare PA15123
 View more versions of this product
Catalog No. 45-001-242
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Steady state Fluorescence, FP and FRET
    Product Type Cy5
    Quantity 3 x 0.5mg,3 x 0.5mg
    Product Name Cy5
  • Description & Specifications
Related Products
Product Content Feedback