Learn More
GE Healthcare SEPHACRYL™ S-1000 Gel Filtration Media
For fast, large-scale purification of plasmid DNA by gel filtration
Manufacturer:  GE Healthcare 17047601
Catalog No. 45-000-084
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Super-fine medium yields highly pure plasmid DNA with no need for ultracentrifugation
  • Heat stability allows sterilization by autoclaving
  • Exceptional flow properties permit columns to be packed and equilibrated quickly and easily
  • Can be used in aqueous buffer systems (pH 2 to 11), concentrated urea or in guanidine hydrochloride, as well as a number of organic solvents
Specifications
Specifications
750 mL
40cm/hr.
Solid Spherical Bead
For the purification of DNA up to 20 000 base pairs, dextrans up to 108 molecular weight, and spherical particles up to 400 nm.
60cm
2 to 11
DNA and large Polysaccharides, Proteoglycans and small particles like membrane-bound vesicles and viruses
4 to 30°C, 20% Ethanol
Sephacryl S-1000 SF, 750 mL
2.6cm
23.62 in.
40 to 105μm
Allyl Dextran and N, N'-methylenebisacrylamide