Learn More
GE Healthcare Sephadex™ Ion Exchange Media
Separate molecules over broad molecular weight and pH ranges
Manufacturer:  GE Healthcare 17024001
 View more versions of this product
Catalog No. 45-000-047
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
100g
<45cm/hr.
Sephadex cation exchange media
Separating and purifying Proteins, Polypeptides, Nucleic Acids and other charged biomolecules by using column or batch techniques
5 to 15cm
SP-Sephadex C-50
2 to 12
30,000 to 100,000
4° to 25°C
Column or Batch Techniques
Solid Spherical Bead
2 to 5.9 in.
Sulphopropyl
40 to 120μm in dry form
Cross-linked Dextran