Learn More
GE Healthcare Sepharose™ Agarose Gel Filtration Media Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  GE Healthcare 17013001
Catalog No. 45-000-030
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Specifications
1L
<10cm/hr.
Solid Bead
60 to 200μm
2% Agarose
4° to 30°C
Sepharose 2B Gel filtration media
Allow characterization or cleaning up of samples containing components of diverse molecular weight
4 to 9