Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva (Formerly GE Healthcare Life Sciences) SOURCE™ 15Q

Strong anion exchanger with high chemical stability for wide range of working conditions with good resistance to cleaning conditions at high pH

$385.50 - $1532.00

Specifications

Buffer 20mM N-methylpiperazine, 20mM Piperazine, 20mM Bis-Tris, 20mM Triethanolamine, 20mM Tris, 20mM Diethanolamine, 20mM Ethanolamine, Glycine, 20mM 1,3-diamino-propane, 20mM Piperidine
Product Type Source 15Q chromato graphy media
Flow Rate 1800cm/hr.
For Use With (Application) Polishing steps in both research and industrial applications
Height (English) 3.93 in.
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
45-002-027
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
17094720
10mL
Each for $385.50
Add to cart
 
45-002-028
View Documents
Cytiva (Formerly GE Healthcare Life Sciences)
17094701
50mL
Each for $1,532.00
Add to cart
 
Description

Description

  • Bioprocess medium
  • Ligand: quaternary ammonium
  • Average particle size: 15µm
  • Matrix: polystyrene/divinylbenzene
  • pH stability working range: 2 to 12
  • For polishing steps in both research and industrial applications
  • Available in 10mL and 50mL bottles

Specifications

Specifications

20mM N-methylpiperazine, 20mM Piperazine, 20mM Bis-Tris, 20mM Triethanolamine, 20mM Tris, 20mM Diethanolamine, 20mM Ethanolamine, Glycine, 20mM 1,3-diamino-propane, 20mM Piperidine
1800cm/hr.
3.93 in.
Quaternary Ammonium
15μm
4° to 30°C
Source 15Q chromato graphy media
Polishing steps in both research and industrial applications
100mm
5 bar
2 to 12
Polystyrene/Divinylbenzene
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit