Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Standard Format HE33 Mini Submarine Unit and Accessories

Enhanced passive cooling system allows running gels at high voltages for speed and resolution

Manufacturer:  GE Healthcare 80605264

 View more versions of this product

Catalog No. 45-001-852

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Includes: Basic HE 33 Mini Submarine Unit:
Gel running tray, gel casting tray and bubble level

Complete HE 33 Mini Submarine Unit:
Basic unit, 1.5mm thick 8-well comb and comb back

Description

Description

  • Coolant-insulated buffer chamber runs 7 x 10cm agarose gels in <20 minutes
  • Convenient, leak-proof casting trays provide fast gel casting without tape
  • UV-transparent gel tray for convenient gel handling and viewing
  • Combs allow up to 32 samples per run
  • Molded base design to be filled with common coolant such as 50% ethylene glycol

Order Info

For Basic unit, combs and comb back should be ordered separately

Specifications

Specifications

500 mA
5.2 x 9.5 x 2.8 in. (13 x 24 x 7 cm)
7 cm
3.9 in.
10 cm
Up to 32
50°C
8.4 oz.
15 w
9.5 in.
24 cm
EN61010-1, UL3101-1, CSA C22.2 1010.1, CE
2.8 in.
Gel running tray, gel casting tray and bubble level
5.2 in.
13 cm
122°F
500 V
250 mL
2.7 in.
7 cm
Mini Submarine Unit (Basic)
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit