Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva (Formerly GE Healthcare Life Sciences) StrepTactin™ Sepharose™ High Performance Medium

Mild purification conditions preserve antigenicity and function. Cytiva StrepTactin Sepharose High Performance media is used for robust, high resolution purification of Strep II-tagged proteins.

Manufacturer:  Cytiva (Formerly GE Healthcare Life Sciences) 28935599

 View more versions of this product

Catalog No. 45-001-585


Add to cart

Description

Description

  • Medium with high binding capacity yields high resolution, sharp concentrated peaks, and reproducible purifications
  • Physiological conditions and mild elution preserves the activity of the target protein
  • Compatible with a wide range of reducing agents, detergents, denaturants, and other additives
  • Easily scaled up, and easily regenerated with 0.5M NaOH
  • Convenient use with a peristaltic pump or chromatography system

  • Matrix: rigid, highly cross-linked 6% agarose
  • Ave. particle size: 34µm
  • Dynamic binding capacity (protein dependent): approx. 6mg Strep(II)-tagged protein/mL medium
  • Compatibility: stable in all commonly used buffers, 0.5M NaOH (regeneration and cleaning), reducing agents, denaturants, and detergents
  • pH stability: >7

Specifications

Specifications

StrepTactin Sepharose HP media
Purification of Strep (II) -tagged recombinant Proteins to high purities in concentrated forms and small volumes by using with perstaltic pump or chromato graphy system
StrepTactin (∼5mg StrepTactin/mL medium)
34μm
2° to 8°C
10mL
<300 cm/hr.
7
6% Highly Cross-linked Agarose
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit