Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Cytiva StrepTrap™ HP Prepacked Columns
Purify Strep(II)-tagged recombinant proteins by affinity chromatography. Cytiva Life Sciences™ StrepTrap™ HP Prepacked Columns are packed with a robust and stable Sepharose™ based medium which results in high resolution, sharp concentrated peaks, and reproducible purifications. The binding capacity is also high.
Supplier: Cytiva 29048653
Description
- Prepacked with StrepTactin Sepharose High Performance
- Binding capacity Approx. 6 mg Strep II-tagged protein/ml medium
- Maximum pressure: 3 bar
- Column material: polypropylene
- Compatible with a wide range of reducing agents, detergents, denaturants, and other additives
- Generated with 0.5M NaOH
- 1 and 5mL columns
Media
- Ligand: Streptactin
- Matrix: 6% cross-linked agarose
- Average particle size: 34μm
- pH stability working range: >7
1mL columns
- Bed dimensions: 7 x 25mm
- Flow velocity: <4mL/min.
5mL columns
- Bed dimensions: 16 x 25mm
- Flow velocity: <20mL/min.
Specifications
StrepTactin Sepharose HP prepacked column | |
0.27 in. | |
1 x 1mL | |
25mm | |
Polypropylene | |
3 bar | |
34μm | |
2° to 8°C | |
Purification of Strep (II) -tagged recombinant Proteins by affinity chromato graphy |
1mL | |
7mm | |
0.98 in. | |
<4mL/min. | |
StrepTactin (∼5mg/mL medium) | |
7 | |
6% Highly Cross-linked Agarose | |
Ion Exchange Column |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction