Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva StrepTrap™ HP Prepacked Columns

Purify Strep(II)-tagged recombinant proteins by affinity chromatography. Cytiva Life Sciences™ StrepTrap™ HP Prepacked Columns are packed with a robust and stable Sepharose™ based medium which results in high resolution, sharp concentrated peaks, and reproducible purifications. The binding capacity is also high.

Supplier:  Cytiva 28907548

 View more versions of this product

Catalog No. 45-001-521

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.


Description

Description

  • Prepacked with StrepTactin Sepharose High Performance
  • Binding capacity Approx. 6 mg Strep II-tagged protein/ml medium
  • Maximum pressure: 3 bar
  • Column material: polypropylene
  • Compatible with a wide range of reducing agents, detergents, denaturants, and other additives
  • Generated with 0.5M NaOH
  • 1 and 5mL columns

Media
  • Ligand: Streptactin
  • Matrix: 6% cross-linked agarose
  • Average particle size: 34μm
  • pH stability working range: >7

1mL columns

  • Bed dimensions: 7 x 25mm
  • Flow velocity: <4mL/min.

5mL columns
  • Bed dimensions: 16 x 25mm
  • Flow velocity: <20mL/min.

Specifications

Specifications

StrepTactin Sepharose HP prepacked column
0.62 in.
5 x 5mL
25mm
Polypropylene
3 bar
34μm
4° to 8°C
Purification of Strep (II) -tagged recombinant Proteins by affinity chromato graphy
5mL
16mm
0.98 in.
<20mL/min.
StrepTactin (∼5mg/mL medium)
7
6% Highly Cross-linked Agarose
Ion Exchange Column
Product Suggestions

Product Suggestions

SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit