Learn More
GE Healthcare Superdex™ Peptide 3.2/300 Prepacked Precision Column
For high-resolution gel filtration of peptides and small biomolecules with molecular weights between 100 and 7000 in micropreparative scale
Manufacturer:  GE Healthcare 29036231
Catalog No. 45-000-652
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Prepacked with Superdex Peptide media
  • Spherical composite of cross-linked agarose and dextran
  • Supplied with two fingertight 1/16 in. male connectors for connection
  • For sensitive and high resolution gel filtration of proteins, peptides, polynucleotides and other biomolecules
  • Used for polishing step in purification process
  • With new hardware design for direct connection to Ä;KTATM systems
Specifications
Specifications
0.01mL/min. to 0.15mL/min.
Borosilicate Glass
13 to 15μm