Learn More
GE Healthcare Superose™ 12 3.2/300 Prepacked Precision Column
The Precision Column PC 3.2/30 (2.4 ml), prepacked with Superose12, is designed for sensitive and highly resolving gel filtration separations of biomolecules in the micropreparative scale. Superose 12 has a broad fractionation range for molecular weights between 1000 and 300 000.
Manufacturer:  GE Healthcare 29036225
Catalog No. 45-000-654
Add to cart
Description
Description
  • Achieve high-resolution separations across exceptionally broad molecular-weight ranges with Superose gel filtration medium.
  • Precision columns (PC) for micropurification and analysis: sample volumes of 4 - 50 µl.
  • Low ionic strength eluents (<0.05 M) can utilize a hydrophobic interaction component to alter the selectivity of Superose for some lipids, peptides, and small aromatic compounds.
  • New hardware design for convenient direct connection to high performance chromatography systems such as ÄKTA design.
Specifications
Specifications
4 to 30°C
2.4 mL
Borosilicate Glass
9 to 13μm
0.01 mL/min-0.10 mL/min
Superose 12 3.2/300
24 bar [2.4MPa] (350 psi)