GE Healthcare Taq DNA Polymerase (cloned) Available on GSA/VA Contract for Federal Government customers only.

For PCR and other procedures requiring DNA polymerase activity at elevated temperatures

Manufacturer: GE Healthcare 27079805

 View more versions of this product

Catalog No. 46-001-009

  Check Availability
Add to Cart

    Specifications

    Color Colorless
    For Use With (Application) For PCR and other procedures requiring DNA Polymerase activity at elevated temperatures
    Purity Free from detectable nonspecific nuclease and Taq I restriction endonuclease activities.
    Quantity 4 x 250U
    Storage Requirements -20°C
    Concentration 5000U/mL,5000U/mL
    pH 7.5
    Includes 100mM Tris HCl, pH 9, 15mM MgCl2 and 500mM KCl

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit