Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare TE 22 Mini Tank Transfer Unit Available on GSA/VA Contract for Federal Government customers only.

Performs protein and nucleic acid transfers for Western, Southern, and Northern blots in less than one hour

Manufacturer:  GE Healthcare 80620426

Catalog No. 45-001-953


Add to cart

Includes: Four gel cassettes; eight foam sponges, 3mm thick; four foam sponges, 6mm thick; and 25 sheets of blotter paper

Description

Description

  • Transfers four mini-gels up to 9 x 10cm in size in about 1L of buffer
  • Built-in alumina-covered heat exchanger dissipates heat generated by tank electrotransfer; typical temperature increase: ≤5°C
  • 80cm of wire in seven parallel strands on each electrode panel delivers a uniform electrical field
  • Hinged, color-coded cassettes allow quick setup of transfers
  • Replacement electrode panel, sponges, and gel cassettes available separately

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Specifications

Specifications

500 mA
6.5 in.
Four gel cassettes, eight 3mm thick Foam sponges, four 6mm thick Foam sponges and 25 sheets of blotter Paper, Transphor tank
100 V
1000 mL
5.5 in.
TE 22 Mini Tank Transfer Unit
EN61010-1, CE 73/23/EEC (LV directive)
16.5 cm
Transfers four mini-gels up to 9 x 10cm
33.81 oz.
50 w
14 cm
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit