Learn More
GE Healthcare TE 62 Transfer Unit
Efficiently transfers proteins, DNA, or RNA from four standard polyacrylamide or agarose gels up to 15 x 21cm in size, or from 16 mini-gels
Manufacturer:  GE Healthcare 80620958
Catalog No. 45-001-962
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Safety lid with power cables; 4 gel cassettes; 8 foam sponges, 3mm thick; 4 foam sponges, 6mm thick; 25 sheets of blotter paper. Replacement gel cassettes, electrode panels and foam sponges available.
Description
Description
  • Hinged cassettes, molded of tough, glass-filled polyethersulfone for high durability, provide equal pressure across the gel
  • Cassettes load easily, with even contact between the gel and transfer membrane
  • Integral heat exchanger serves as a buffer cooling system
  • Safe, sturdy electrode panels lock automatically in place
  • Evenly spaced vertical platinum wire on panel and 10cm spacing between panels promote efficient transfer
Specifications
Specifications
Black, Gray
16.5 cm
39.2°F to 104°F
200 w
28 cm
6.49 in.
Safety lid with power cables, 4 gel cassettes, 8 Foam sponges 3mm thick, 4 Foam sponges 6mm thick, 25 sheets of blotter Paper
4°C to 40°C
11.02 in.
Blotting System