Learn More
GE Healthcare Thiopropyl Sepharose™ 6B Gel Available on GSA/VA Contract for Federal Government customers only.
Used for reversible immobilization of molecules containing thiol groups under mild conditions. Thiopropyl Sepharose™ 6B Gels are optimized for immobilization of small biomolecules.
Manufacturer:  GE Healthcare 17042001
Catalog No. 45-000-065
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Small hydroxypropyl spacer arm associated with Sepharose 6B matrix
  • Gel also reacts with heavy metal ions and with alkyl and aryl halides
  • Additional reactions occur with compounds containing C=O, C=C and N=N bonds
  • Coupling is reversible with low molecular weight thiols
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Used for reversible immobilization of molecules containing Thiol groups under mild conditions, optimized for immobilization of small biomolecules
20μm
6% agarose
90μm
2° to 8°C
Thiolated
Reversible
6% Agarose
15g