GE Healthcare TiO2 Mag Sepharose™

Designed for enrichment of phosphopeptides from tryptic digested protein samples. GE Healthcare TiO2 Mag Sepharose magnetic beads are based on titanium dioxide chromatography and are designed to simplify capture and enrichment of phosphopeptides.

Manufacturer: GE Healthcare 28951377

 View more versions of this product

Catalog No. 45-002-175

  Check Availability
Add to Cart

    Specifications

    Storage Requirements 4° to 30°C (use at room temperature)
    Storage Buffer 20% ethanol storage solution
    For Use With (Application) Used for selective capture and enrichment of Phosphopeptides, for biomarker research
    Ligand TiO2
    Matrix Paramagnetic spherical, highly cross linked agarose particles
    Particle Size 37 to 100μm
    Bead Type TiO2 Mag Sepharose
    Quantity 4 x 500μL

    TiO2 Mag Sepharose has high affinity for phosphopeptides and is an efficient method for enrichment of phosphopeptides from complex samples. The magnetic bead format improves ease-of-use in sample preparation and has excellent properties for small-scale experiments. The high density beads allows rapid capture by magnetic devices with high visibility to ensure reliable collection of all bound target peptides.

    • Magnetic beads for selective capture and enrichment of phosphopeptides
    • Easy to use - visible and dense magnetic beads
    • Simple protocol with elution conditions optimized for MS analysis
    • Parallel sample handling when used with MagRack 6 separation tool - process up to six samples in less than one hour

     

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit