Learn More
GE Healthcare TiO2 Mag Sepharose™ Available on GSA/VA Contract for Federal Government customers only.
Designed for enrichment of phosphopeptides from tryptic digested protein samples. GE Healthcare TiO2 Mag Sepharose magnetic beads are based on titanium dioxide chromatography and are designed to simplify capture and enrichment of phosphopeptides.
Manufacturer:  GE Healthcare 28944010
 View more versions of this product
Catalog No. 45-002-158
Add to cart
Description
Description
TiO2 Mag Sepharose has high affinity for phosphopeptides and is an efficient method for enrichment of phosphopeptides from complex samples. The magnetic bead format improves ease-of-use in sample preparation and has excellent properties for small-scale experiments. The high density beads allows rapid capture by magnetic devices with high visibility to ensure reliable collection of all bound target peptides.
  • Magnetic beads for selective capture and enrichment of phosphopeptides
  • Easy to use - visible and dense magnetic beads
  • Simple protocol with elution conditions optimized for MS analysis
  • Parallel sample handling when used with MagRack 6 separation tool - process up to six samples in less than one hour
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
TiO2 Mag Sepharose
TiO2
37 to 100μm
4° to 30°C (use at room temperature)
Used for selective capture and enrichment of Phosphopeptides, for biomarker research
Paramagnetic spherical, highly cross linked agarose particles
500μL
20% ethanol storage solution