Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GE Healthcare Tricorn™ Superdex™ Prepacked Gel Filtration Columns Available on GSA/VA Contract for Federal Government customers only.

Use for laboratory-scale separations and intermediate purification based on different sizes of proteins, peptides, and other biomolecules. GE Healthcare Life Sciences™Tricorn™ Superdex™ Prepacked Gel Filtration Columns are perfect for high-resolution applications.

Manufacturer:  GE Healthcare 28920504

 View more versions of this product

Catalog No. 45-002-907


  Check Availability
Add to cart

Specifications

Description

  • Fast gel filtration involving separation based on three molecular weight ranges from 100 to 600000 M
  • Spherical composite of cross-linked agarose and dextran matrix
  • Comes with two different bed heights of 150 and 300mm allowing greater flexibility to suit different analytical needs
  • Available with three different media: Superdex Peptide, Superdex 75, and Superdex 200

Superdex Peptide:

  • Bed dimension: 10 x 300mm
  • Bed volume: 24mL
  • Flow velocity: 0.15 to 0.6mL/min.
  • Pressure max.: 18 bar (261psi)
  • Separation range: 100 to 7000 (peptides)
  • pH Stability working range: 1 to 14
  • Column I.D.: 10mm
  • Bed height with one adaptor:     279 to 309mm
  • Bed height with two adaptors: 245 to 306mm
Superdex 200:
  • Bed dimension: 5 x 150mm and 10 x 300mm
  • Bed volume: 3 and 24mL
  • Flow velocity: 0.15 to 0.6mL/min. and 0.25 to 0.75mL/min.
  • Pressure max.: 15 bar (217psi)
  • Separation range: 10,000 to 600,000 (globular peptides)
  • pH Stability working range: 3 to 12
  • Column I.D.: 5 and 10mm
  • Bed height with one adaptor:     135 to 159mm (3mL column)
  • Bed height with two adaptors: 108 to 156mm (3mL column)
Superdex 75:
  • Bed dimension: 5 x 150mm and 10 x 300mm
  • Bed volume: 3 and 24mL
  • Flow velocity: 0.15 to 0.6mL/min.
  • Pressure max.: 18 bar (261psi)
  • Separation range: 3000 up to 70,000 (globular proteins)
  • pH Stability working range: 3 to 12
  • Column I.D.: 5 and 10 mm
  • Bed height with one adaptor: 108 to 156mm (3mL column)
  • Bed height with two adaptors: 135 to 159mm (3mL column)

Description & Specifications

Specifications

1M Acetic Acid, 8M Urea, 6M Guanidine Hydrochloride, 30% Isopropanol, 70% Ethanol, 1M NaOH
0.19 in.
Superdex™ 75 prepacked column
High-resolution, semipreparative and analytical separations of biomolecules
150mm
Valco fittings for AKTA™ design systems and are supplied with M6 connecto rs for FPLC System
18 bar
13μm
4° to 30°C
3mL
5mm
Each
5.9 in.
0.15 to 0.6mL/min.
Borosilicate Glass
1 to 14
Spherical composite of cross-linked Agarose and Dextran
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit