GE Healthcare Tricorn™ Superose™ 6 and Superose 12 Prepacked Gel Filtration Columns

Designed for high performance, laboratory scale separations of proteins and other biomolecules based on size.  GE Healthcare Tricorn™ Superose™ 6 and Superose 12 Prepacked Gel Filtration Columns  delivers quick and reproducible separations with high resolution.

Manufacturer: GE Healthcare 17517201

 View more versions of this product

Catalog No. 45-002-901

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Buffer 1M Acetic Acid, 8M Urea, 6M Guanidine Hydrochloride, 1% SDS, Organic Solvents, 0.5M NaOH (for cleaning in-place)
    Quantity Each
    For Use With (Application) High performance gel filtration with a broad protein fractionation range of 5000 to 5,000,000
    Height (English) 11.81 in.
    Height (Metric) 300mm
    Flow Rate 0.3 to 0.5mL/min.
    Includes Valco fittings for AKTA™ design systems and are supplied with M6 connectors for FPLC System
    Material Borosilicate Glass
    Max. Pressure 217psi
    pH Range 3 to 12
    Particle Size 13μm
    Support Type Cross-linked Agarose
    Storage Requirements 4° to 30°C
    Bed Volume 24mL
    Diameter (English) Inner 0.39 in.
    Diameter (Metric) Inner 10mm
    Product Type Superose 6 prepacked column

     

    • Covers wide separation range of molecular weights from 1000 up to 5million MHighly cross-linked agarose matrixSmall average bead size of 13µm (Superose 6) and 11µm (Superose 12) are designed for laboratory-scale separations with good resolving capability
    • Durable, high-performance glass columns for reliable and reproducible separations
    • Available with two different media - Superose 6 or Superose 12

       

       

    • Bed volume: 24mL
    • Bed dimension: 10 x 300mm
    • Pressure max.: 15 bar (217psi)

    • Media

    • pH stability working range: 3 to 12
    • pH stability cleaning: 1 to 14

    • Column

    • Column I.D.: 10mm
    • Bed support: filter
    • Material (bed support): polypropylene
    • Material (column hardware): borosilicate glass, PEEK, PP, EPDM and PFR
    • Material (column tube): borosilicate glass
    • Bed height with one adaptor: 279 to 309mm
    • Bed height with two adaptors: 245 to 306mm
    •  

      Superose 6 Media:

    • Average partcle size: 13µm
    • Gel filtration separations over a very broad molecular weight rang from 5000 to 5,000,000 (globular proteins)
    • Flow rate: 0.3 to 0.5mL/min.

    • Superose 12 Media:

    • Average partcle size: 11µm
    • Gel filtration separations over a very broad molecular weight rang from 1000 to 300,000 (globular proteins)
    • Flow rate of 0.5 to 1mL/min.
    •  

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit