GE Healthcare Ultrospec™ 2100 pro UV/Visible Spectrophotometers Available on GSA/VA Contract for Federal Government customers only.

Simple-to-use instrument for nucleic acid measurements and enzyme kinetics

Manufacturer: GE Healthcare 80211221

 View more versions of this product

Catalog No. 45-001-800

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type UV-visible
    Warranty 3 Year
    Electrical Requirements 100 to 240V 50/60Hz
    For Use With (Application) Nucleic acid measurements and enzyme kinetics
    Accuracy (Photometric) ±0.5% or ±0.003A to 3.000A at 54nm
    Certifications/Compliance CE89/336/EEC (EMCdirective); CE73/23/EEC (LVdirective); EN-61010-1 (IEC1010-1)
    Width (English) 8.26 in.
    Width (Metric) 21cm
    Depth (English) 5.5 in.
    Depth (Metric) 14cm
    Detector Type Two Silicon Photodiodes
    Hertz 50/60Hz
    Lamp Xenon
    Voltage 100 to 240V
    Spectral Bandwidth >3nm
    Wavelength Accuracy ±1nm
    Wavelength Range 190 to 900nm
    Weight (English) 28 lb.
    Weight (Metric) 13kg
    Height (English) 3.14 in.
    Height (Metric) 8cm
    Scan Speed 3000nm/min.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit