Learn More
GE Healthcare Variable Pathlength Cell Holder
For use with Ultrospec 7000/8000/9000.
Manufacturer:  GE Healthcare 80710005
Catalog No. 45-002-320
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • Holds 1, 2, 5, 10, 20, 50 and 100mm pathlength cells
  • Includes one thumb screw
Specifications
Specifications
Ultrospec 7000/8000/9000
1, 2, 5, 10, 20, 50 and 100mm pathlength cells
One thumb screw
Cell Holder