Learn More
Cytiva Vivaspin™ 2 Sample Concentrators
Use for one-step, non-denaturing sample concentration in a single tube for minimal sample handling and reduced sample loss. Cytiva Life Vivaspin™ 2 Sample Concentrators cater to scientists working with sample volumes ranging from 100 μl to 20 ml, with a range of MWCO values from Mr 3 000 to 100 000.
$257.50
Specifications
Material (Membrane) | PES |
---|---|
Filtration Area | 1.2 cm2 |
Quantity | 25/Pk. |
Volume (Metric) | 400 μL to 2 mL |
Length (Metric) | 126 mm |
Catalog Number | Mfr. No. | MWCO | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | MWCO | Price | Quantity & Availability | |||||
45-001-568
|
Cytiva
28932248 |
30 kDa |
Pack of 25 for $257.50
|
|
|||||
45-001-566
|
Cytiva
28932245 |
5 kDa |
Pack of 25 for $257.50
|
|
|||||
45-001-569
|
Cytiva
28932257 |
50 kDa |
Pack of 25 for $257.50
|
|
|||||
45-001-567
|
Cytiva
28932247 |
10 kDa |
Pack of 25 for $257.50
|
|
|||||
45-001-565
|
Cytiva
28932240 |
3 kDa |
Pack of 25 for $257.50
|
|
|||||
45-001-570
|
Cytiva
28932258 |
100 kDa |
Pack of 25 for $257.50
|
|
|||||
Description
Up to 30-fold concentration of the sample can be achieved with recovery of the target molecule typically exceeding 95%. Patented dead-stop technology ensures that samples cannot be concentrated to dryness and enables direct concentrate recovery.
- Accept sample volumes from 100μLto 20mL, with a range of molecular weight cutoff values from Mr3000 to 100,000
- Vertical polyethersulfone semi-permeable membrane separating upper and lower chamber resists blockage and tolerates high flow rate
- Centrifugation forces solvent through membrane, leaving concentrated sample in the upper chamber
- Compatible pH range: 1 to 9
- Active membrane area: 1.2cm2
- Vivaspin 2 offers easy, contact-free storage by reverse spinning the concentrate into the recovery cap
For maximum recovery, select a MWCO at least 50% smaller than the molecular size of the species of interest.
Specifications
PES | |
25/Pk. | |
126 mm |
1.2 cm2 | |
400 μL to 2 mL | |
17 mm |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.