Learn More
GE Healthcare Water Thermostatted Single Cell Holder
Works with water circulators to achieve constant temperatures in 5° to 80°C range.
Manufacturer:  GE Healthcare 80710003
Catalog No. 45-002-321
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
  • 10mm pathlength
  • One thumb screw
  • For Ultrospec 7000/8000/9000
Specifications
Specifications
Ultrospec 7000/8000/9000
One thumb screw
Water