Learn More
Cytiva (Formerly GE Healthcare Life Sciences) Yeast Protein Extraction Buffer Kit
Compatible with any downstream application including enzymes assays, chromatography procedures, and gel electrophoresis applications
Manufacturer:  Cytiva (Formerly GE Healthcare Life Sciences) 28944045
Catalog No. 45-002-157
Add to cart
Includes:
  • 100mL Yeast protein extraction buffer
  • 15mL Yeast suspension buffer
  • 2 x 0.5mL Longlife Zymolyase
Description
Description
  • Mild detergent-based cell lysis method
  • For total extraction of soluble proteins from yeast cells
  • Eliminates the need for glass beads
  • Suitable for preparation of approximately 10mL yeast cell pellets suspension as single or as multiple smaller preps
Specifications
Specifications
1 Piece
Total extraction of soluble Proteins from yeast cells, downstream application including Enzymes assays, chromato graphy procedures and Gel electrophoresis applications
Longlife Zymolyase below -18°C and other kit components at 2–8°C Stable for 1 year from manufacturing date when stored and used as recommended
Yeast Protein Extraction Buffer Kit
100 mL Yeast protein extraction buffer , 15 mL Yeast sSuspension buffer , 2 × 0.5 mL Longlife Zymolyase (1500 U/ml)