Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GEF-H1 Mouse anti-Human, Mouse, Rat, Biotin, Clone: 336, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP243635B

Catalog No. NBP243635B


Add to cart

Description

Description

GEF-H1 Monoclonal antibody specifically detects GEF-H1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

GEF-H1
336
Western Blot, Immunocytochemistry/Immunofluorescence
Purified
ARHGEF2
Recombinant protein encompassing a sequence within the C-terminus region of human GEF-H1. The exact sequence is proprietary.
Protein G purified
Cell Cycle and Replication
Primary
9181
Western Blot, Immunocytochemistry, Immunofluorescence
Biotin
PBS with 0.05% Sodium Azide
GEF, GEFH1, GEF-H1DKFZp547P1516, Guanine nucleotide exchange factor H1, KIAA0651DKFZp547L106, LFP40FLJ45440, Microtubule-regulated Rho-GEF, P40, Proliferating cell nucleolar antigen p40, rho guanine nucleotide exchange factor 2, Rho/Rac guanine nucleotide exchange factor (GEF) 2, Rho/Rac guanine nucleotide exchange factor 2
Mouse
IgG1
0.1 ml
Store at 4C in the dark.
Monoclonal
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit