Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
GEF-H1 Mouse anti-Human, Unconjugated, Clone: 3C5, MilliporeSigma™
Mouse Monoclonal Antibody
$184.00 - $463.00
Specifications
Antigen | GEF-H1 |
---|---|
Clone | 3C5 |
Applications | Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABC10962MI
|
MilliporeSigma
MABC109625UG |
25 μg |
Each for $184.00
|
|
|||||
MABC10961MI
|
MilliporeSigma
MABC1096100UG |
100 μg |
Each for $463.00
|
|
|||||
Description
GEF-H1 Monoclonal antibody specifically detects GEF-H1 in Human samples. It is validated for Western Blotting.Specifications
GEF-H1 | |
Western Blot | |
Unconjugated | |
Mouse | |
Human | |
Rho guanine nucleotide exchange factor 2;Guanine nucleotide exchange factor H1;Microtubule-regulated Rho-GEF;Proliferating cell nucleolar antigen p40 | |
A linear peptide from the N-terminal region of human GEF-H1 | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼111 kDa observed; 111.54 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
3C5 | |
Monoclonal | |
Purified | |
Apoptosis and Cancer | |
NP_001155855 | |
ARHGEF2, KIAA0651, LFP40 | |
IgG1 κ | |
Protein G Purified | |
Clone 3C5 is a mouse monoclonal antibody that specifically detects human Rho guanine nucleotide exchange factor 2 (GEF-H1). |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title