Learn More
Aviva Systems Biology Corp GEFT Antibody - C-terminal region (ARP53438_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP53438_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against GEFT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human GEFT. Alternate names: p63RhoGEF, GEFTApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-61024