Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

GEFT Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.

Rabbit Polyclonal Antibody

Manufacturer:  Proteintech Group IncSupplier Diversity Partner 148391AP

Catalog No. 50-172-9767


Add to cart

Description

Description

The Rho family of small GTPases act as molecular switches to control a wide range of cellular processes. Guanine nucleotide exchange factors (GEFs), like GEFT, activate Rho GTPases by accelerating GDP/GTP exchange. The Rho family of small GTPases act as molecular switches to control a wide range of cellular processes. Guanine nucleotide exchange factors (GEFs), like GEFT, activate Rho GTPases by accelerating GDP/GTP exchange (Souchet et al., 2002 ).
Specifications

Specifications

GEFT
0.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q6P720, Q86VW2, Q9CWR0
ARHGEF25, GEFT, p63RhoGEF, RhoA/RAC/CDC42 exchange factor
Rabbit
IgG
150 μL
-20°C
Primary
115557, 314904, 52666
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Unconjugated
ARHGEF25
Liquid
Arhgef25
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat
Documents
Provide Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit